2,6-Dichloro-4-iodopyridine, Sigma-Aldrich®
Catalog # SIAL654078-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:273.89 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:98027-84-0
Formula:C₅H₂CI₂IN
MDL Number:MFCD07368400