
2',3'-Dideoxyadenosine ≥98.0% (by HPLC, titration analysis)
TCIAD3065-100MG
: TCI
:
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:186.0 to 190.0 deg-C
- Specific rotation [a]20/D:-27.0 to -34.0 deg (C=1, H2O)
:
:14,03101
:235.24 g/mol
:223-853-2
:GHS08
:4097-22-7
:C₁₀H₁₃N₅O₂
:MFCD00010534