5,5-Diphenylhydantoin
Catalog # PROOICRS0387
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:252.27 g/mol
EINECS:200-328-6
CAS Number:57-41-0
Formula:C₁₅H₁₂N₂O₂
MDL Number:MFCD00005264