Order Entry
Export
ContactUsLinkComponent
Cycloheptanol ≥98.0% (by GC)
Cycloheptanol ≥98.0% (by GC)
Catalog # TCIAC0678-25ML
Supplier:  TCI
CAS Number:  
undefined
Cycloheptanol ≥98.0% (by GC)
Catalog # TCIAC0678-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :114.19 g/mol
Boiling Pt:185°C
Melting Pt:2 °C
Density:0,9554 g/cm³ (20 °C)
Flash Pt:71°C
Storage Temperature:Ambient
EINECS:207-936-0
CAS Number:502-41-0
Formula:C₇H₁₄O
MDL Number:MFCD00004150