Order Entry
Export
Orders LinkContactUsLinkComponent
1-Hexadecanol

1-Hexadecanol

Glass bottle for solids, 1 kg
Catalog # ROTH5762.1
Supplier:   Roth Carl
1-Hexadecanol
Catalog # ROTH5762.1
Supplier:  Roth Carl
CAS Number:  36653-82-4
1-Hexadecanol

Glass bottle for solids, 1 kg

CAS Number:  36653-82-4
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Cetyl alcohol, Palmityl alcohol
  • CAS Number:36653-82-4
  • Formula:H₃C(CH₂)₁₄CH₂OH
  • MDL Number:MFCD00004760
  • Molecular Weight:242.45 g/mol
  • Merck Index:13,02037
  • Boiling Pt:305…330 °C (1013 hPa)
  • Melting Pt:46…52 °C
  • Density:0.81 g/cm³ (20 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:253-149-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications