2,2'-Dibromobiphenyl (PBB No. 4)
Catalog # EHERC21000400
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:312 g/mol
CAS Number:13029-09-9
Formula:C₁₂H₈Br₂
MDL Number:MFCD00093707