Ammonium hexafluorosilicate
Glass bottle for solids, 10 g
Catalog # ROTH5587.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Ammonium silicofluoride Ammonium hexafluorosilicate, Ammonium fluorosilicate, Ammomium fluosilicate
- CAS Number:16919-19-0
- Formula:H₈F₆N₂Si
- MDL Number:MFCD00010887
- Molecular Weight:178.15 g/mol
- Merck Index:13,00529
- Density:2.01 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:240-968-3
- UN:2854
- ADR:6.1,III
- :Density: 2.01 g/cm³ (20 °C)
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



