General purpose gloves, KSR®
Supplier: Ansell
About this item
This interlock cotton liner with full PVC coating is ideal for long-wearing light-duty work.
- Light, flexible and comfortable: ideal for long wearing times
- Excellent grip for handling small and difficult objects
- Slip-on cuff style
Suitable applications include maintenance, shipping and receiving, product inspection, packaging, working with tools and handling aluminium profiles.
Certifications: EN 388:2016 (2121A); Cat. II
5 Options Available Below
Product Details & Documents
This interlock cotton liner with full PVC coating is ideal for long-wearing light-duty work.
- Light, flexible and comfortable: ideal for long wearing times
- Excellent grip for handling small and difficult objects
- Slip-on cuff style
Suitable applications include maintenance, shipping and receiving, product inspection, packaging, working with tools and handling aluminium profiles.
Certifications: EN 388:2016 (2121A); Cat. II
Specifications for Product Family
Specifications
- Catalog No:
- ANSE22-515/7
- ANSE22-515/7.5
- ANSE22-515/8
- ANSE22-515/9
- ANSE22-515/10
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Cotton
- Finish:
- Fully coated
- Fully coated
- Fully coated
- Fully coated
- Fully coated
- Cuff style:
- Slip-on
- Slip-on
- Slip-on
- Slip-on
- Slip-on
- EN norm:
- EN 388:2016 (2121A)
- EN 388:2016 (2121A)
- EN 388:2016 (2121A)
- EN 388:2016 (2121A)
- EN 388:2016 (2121A)
Specifications
Product Family Options
Product Information
- Glove sizeColourLengthPackedAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.ANSE22-515/7Ambidextrous / hand-specificHand-specificDisposable / reusableReusableLiner materialCottonFinishFully coatedCuff styleSlip-onEN normEN 388:2016 (2121A) - Specifications:
More
Specifications
Cat. No.ANSE22-515/7.5Ambidextrous / hand-specificHand-specificDisposable / reusableReusableLiner materialCottonFinishFully coatedCuff styleSlip-onEN normEN 388:2016 (2121A) - Specifications:
More
Specifications
Cat. No.ANSE22-515/8Ambidextrous / hand-specificHand-specificDisposable / reusableReusableLiner materialCottonFinishFully coatedCuff styleSlip-onEN normEN 388:2016 (2121A) - Specifications:
More
Specifications
Cat. No.ANSE22-515/9Ambidextrous / hand-specificHand-specificDisposable / reusableReusableLiner materialCottonFinishFully coatedCuff styleSlip-onEN normEN 388:2016 (2121A) - Specifications:
More
Specifications
Cat. No.ANSE22-515/10Ambidextrous / hand-specificHand-specificDisposable / reusableReusableLiner materialCottonFinishFully coatedCuff styleSlip-onEN normEN 388:2016 (2121A)