2-(Hydroxymethyl)-15-crown-5, Sigma-Aldrich®
Catalog # SIAL388424-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: 2-(Hydroxymethyl)-15-crown-5-ether
:
:250.29 g/mol
:M-Clarity™ quality level = MQ100
EINECS:278-222-4
CAS Number:75507-25-4
Formula:C₁₁H₂₂O₆
MDL Number:MFCD00192215