Order Entry
Export
Orders LinkContactUsLinkComponent
(S)-17-HDoHE

(S)-17-HDoHE

Supplier:   Cayman Chemical
(S)-17-HDoHE
Supplier:  Cayman Chemical
Certificates

4 Options Available Below

Product Details & Documents
  • CAS Number:92693-03-3
  • Formula:C₂₂H₃₂O₃
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 25 µG
    Log in for availability
    Log in for availability
    1 * 25 µG
    1 * 25 µG - 99,00 €
    99,00 €
    1 * 25 µG
       99,00 €

  • 50 µG
    Log in for availability
    Log in for availability
    1 * 50 µG
    1 * 50 µG - 178,00 €
    178,00 €
    1 * 50 µG
       178,00 €

  • 100 µG
    Log in for availability
    Log in for availability
    1 * 100 µG
    1 * 100 µG - 327,00 €
    327,00 €
    1 * 100 µG
       327,00 €

  • 250 µG
    Log in for availability
    Log in for availability
    1 * 250 µG
    1 * 250 µG - 767,00 €
    767,00 €
    1 * 250 µG
       767,00 €