Order Entry
Export
Orders LinkContactUsLinkComponent
(±)-13-HDoHE

(±)-13-HDoHE

Supplier:   Cayman Chemical
(±)-13-HDoHE
Supplier:  Cayman Chemical
Certificates

4 Options Available Below

Product Details & Documents
  • CAS Number:90780-53-3
  • Formula:C₂₂H₃₂O₃
  • UN:1170
  • ADR:3,II
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 25 µG
    Log in for availability
    Log in for availability
    1 * 25 µG
    1 * 25 µG - 81,00 €
    81,00 €
    1 * 25 µG
       81,00 €

  • 50 µG
    Log in for availability
    Log in for availability
    1 * 50 µG
    1 * 50 µG - 152,00 €
    152,00 €
    1 * 50 µG
       152,00 €

  • 100 µG
    Log in for availability
    Log in for availability
    1 * 100 µG
    1 * 100 µG - 285,00 €
    285,00 €
    1 * 100 µG
       285,00 €

  • 250 µG
    Log in for availability
    Log in for availability
    1 * 250 µG
    1 * 250 µG - 672,00 €
    672,00 €
    1 * 250 µG
       672,00 €