Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
(±)-17-HDoHE
(±)-17-HDoHE
(±)-17-HDoHE
Print
(±)-17-HDoHE
Supplier:
Cayman Chemical
(±)-17-HDoHE
Supplier:
Cayman Chemical
Certificates
Expand image
Return to Top
(±)-17-HDoHE
Product Family Options
Add to cart
Jump to:
Product Family Options
About this item
CAS Number:
90780-52-2
Formula:
C₂₂H₃₂O₃
4 Options Available Below
Shop Now
Return Policy
Product Details & Documents
CAS Number:
90780-52-2
Formula:
C₂₂H₃₂O₃
Product Family Options
Product Information
Pk
Availability
Price
CAYM33650-25
25 µG
Log in
for availability
Log in
for availability
1 * 25 µG
1 * 25 µG - 106,00 €
106,00 €
-
+
1 * 25 µG
106,00 €
-
+
CAYM33650-50
50 µG
Log in
for availability
Log in
for availability
1 * 50 µG
1 * 50 µG - 202,00 €
202,00 €
-
+
1 * 50 µG
202,00 €
-
+
CAYM33650-100
100 µG
Log in
for availability
Log in
for availability
1 * 100 µG
1 * 100 µG - 380,00 €
380,00 €
-
+
1 * 100 µG
380,00 €
-
+
CAYM33650-250
250 µG
Log in
for availability
Log in
for availability
1 * 250 µG
1 * 250 µG - 792,00 €
792,00 €
-
+
1 * 250 µG
792,00 €
-
+