Dicyclohexano-18-crown-6 (mixture of isomers)
Catalog # MOLE27631200-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:372.51 g/mol
EINECS:240-216-4
UN:2811
ADR:6.1,III
CAS Number:16069-36-6
Formula:C₂₀H₃₆O₆
MDL Number:MFCD00005099