4'-Nitrobenzo-18-crown-6 (4'-nitrobenzo-1,4,7,10,13,16-hexoxacyclooctadecane)
Catalog # MOLE88364334-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: 18-Nitro-2,3,5,6,8,9,11,12,14,15-decahydrobenzo[b][1,4,7,10,13,16]hexaoxacyclooctadecine
:
:357.35 g/mol
CAS Number:53408-96-1
Formula:C₁₆H₂₃NO₈
MDL Number:MFCD00143207