4,4'-(Hexafluoroisopropylidene)dianiline, Sigma-Aldrich®
Catalog # SIAL368148-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:334.2600 g/mol
CAS Number:1095-78-9
Formula:C₁₅H₁₂F₆N₂
MDL Number:MFCD00039146