
4,4'-(Hexafluoroisopropylidene)dianiline 98%
:1095-78-9
:
:C₁₅H₁₂F₆N₂
:MFCD00039146
:334.2600 g/mol
:195…199 °C
:GHS07
:Ambient
- Appearance:White to light yellow Powder or crystals
- Infrared spectrum:Conforms
- Melting point:195 to 199 °C
- Titration with HClO4:97.5 to 102.5 %