
7-Hydroxycitronellal 97%
ACRO259680250
:
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4460 to 1.4520 (20°C, 589 nm)
- Density:(20°C) 0.916 to 0.924 g/ml
- GC:≥96.0 %
:
:172.2646 g/mol
:241 °C
:104 °C
:203-518-7
:GHS05
:Ambient
:107-75-5
:C₁₀H₂₀O₂
:MFCD00014681