
2,2-Dibromomalonamide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:73003-80-2
:
Formula:C₃H₄Br₂N₂O₂
MDL Number:MFCD00014803
Melting Pt:202…207 °C
:259.89 g/mol
EINECS:277-205-9
:GHS07
Storage Temperature:Ambient