Order Entry
Export
ContactUsLinkComponent
Tetrabutylammonium acetate 1 mol/l in aqueous solution, Sigma-Aldrich®
Tetrabutylammonium acetate 1 mol/l in aqueous solution, Sigma-Aldrich®
Catalog # SIAL401803-50ML
Supplier:  Merck
CAS Number:  
Tetrabutylammonium acetate 1 mol/l in aqueous solution, Sigma-Aldrich®
Catalog # SIAL401803-50ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
 :301.51 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:234-101-8
CAS Number:10534-59-5
Formula:C₁₈H₃₉NO₂
MDL Number:MFCD00043208