Order Entry
Export
ContactUsLinkComponent
2',7'-Dichlorofluorescein
2',7'-Dichlorofluorescein
Catalog # ROTHX866.1
Supplier:  Roth Carl
CAS Number:  
2',7'-Dichlorofluorescein
Catalog # ROTHX866.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :401.20 g/mol
EINECS:200-968-6
CAS Number:76-54-0
Formula:C₂₀H₁₀Cl₂O₅
MDL Number:MFCD00005047