Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Phenylethanethiol
2-Phenylethanethiol, Sigma-Aldrich®
2-Phenylethanethiol, Sigma-Aldrich®
2-Phenylethanethiol, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
2-Phenylethanethiol, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
4410-99-5
:
Formula:
C₈H₁₀S
MDL Number:
MFCD00004891
:
138.2300 g/mol
:
M-Clarity™ quality level = MQ300
EINECS:
224-563-9