Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Dimethylresorcinol
2,5-Dimethylresorcinol
2,5-Dimethylresorcinol
2,5-Dimethylresorcinol
MOLEM77999497
:
Molekula
:
2,5-Dimethylresorcinol
MOLEM77999497
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
138.1638 g/mol
:
207-688-3
:
488-87-9
:
C₈H₁₀O₂
:
MFCD00134473