2-Hydroxycyclohexanone dimer, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
- CAS Number:30282-14-5
- Formula:C₆H₁₀O₂
- MDL Number:MFCD00151157
- Molecular Weight:228.28 g/mol
- ADR:4.1,III
- :M-Clarity™ quality level = MQ100
- UN:1325
2 Options Available Below
Product Details & Documents
- CAS Number:30282-14-5
- Formula:C₆H₁₀O₂
- MDL Number:MFCD00151157
- Molecular Weight:228.28 g/mol
- ADR:4.1,III
- :M-Clarity™ quality level = MQ100
- UN:1325



