2-Hydroxycyclohexanone dimer
Glass bottle for solids, 5 g
Catalog # MOLEM65156295
Supplier: Molekula
Jump to:
Product Details & Documents
- CAS Number:30282-14-5
- Formula:C₆H₁₀O₂
- MDL Number:MFCD00151157
- Molecular Weight:228.28 g/mol
- UN:1325
- ADR:4.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



