Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,6-Dichlororesorcinol
4,6-Dichlororesorcinol, Sigma-Aldrich®
4,6-Dichlororesorcinol, Sigma-Aldrich®
SIAL130982-25G
:
Merck
:
4,6-Dichlororesorcinol, Sigma-Aldrich®
SIAL130982-25G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
179.00 g/mol
:
M-Clarity™ quality level = MQ100
:
205-284-1
:
137-19-9
:
C₆H₄Cl₂O₂
:
MFCD00002274