Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dicyanopyrazine
2,3-Dicyanopyrazine, Sigma-Aldrich®
2,3-Dicyanopyrazine, Sigma-Aldrich®
2,3-Dicyanopyrazine, Sigma-Aldrich®
SIAL399620-5G
:
Merck
:
2,3-Dicyanopyrazine, Sigma-Aldrich®
SIAL399620-5G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
130.1069 g/mol
:
M-Clarity™ quality level = MQ100
:
13481-25-9
:
C₆H₂N₄
:
MFCD00059771