Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Phenylene diisocyanate
1,4-Phenylene diisocyanate, Sigma-Aldrich®
1,4-Phenylene diisocyanate, Sigma-Aldrich®
1,4-Phenylene diisocyanate, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
1,4-Phenylene diisocyanate, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
104-49-4
:
Formula:
C₈H₄N₂O₂
:
M-Clarity™ quality level = MQ100
EINECS:
203-207-6