Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Undecyl β-D-glucopyranoside
Undecyl β-D-glucopyranoside
Undecyl β-D-glucopyranoside
Undecyl β-D-glucopyranoside
Catalog # MOLE72880752-250MG
Supplier:
Molekula
CAS Number:
Expand image
Undecyl β-D-glucopyranoside
Catalog # MOLE72880752-250MG
Supplier:
Molekula
CAS Number:
Specifications
Pk:
250 mg
Seq:
2590
Environmentally Preferable:
Specifications
About this item
Product Details
:
CAS Number:
70005-86-6
Formula:
C₁₇H₃₄O₆
Product Details