4,4-Difluorocyclohexanone, Sigma-Aldrich®
Catalog # SIAL711403-250MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:134.12 g/mol
CAS Number:22515-18-0
Formula:C₆H₈F₂O
MDL Number:MFCD08146635