2-Fluorophenylboronic acid
Glass bottle for solids, 100 g
Catalog # MOLEM18550854
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: Dihydroxy(2-fluorophenyl)borane, 2-Fluorobenzeneboronic acid, (2-Fluorophenyl)metaboric acid, (2-Fluorophenyl)boronic acid
- CAS Number:1993-03-9
- Formula:C₆H₆BFO₂
- MDL Number:MFCD00674013
- Molecular Weight:139.92 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



