Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Luminol (3-Aminophthalhydrazide)
Luminol (3-Aminophthalhydrazide), Sigma-Aldrich®
Luminol (3-Aminophthalhydrazide), Sigma-Aldrich®
Luminol (3-Aminophthalhydrazide), Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Luminol (3-Aminophthalhydrazide), Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
521-31-3
:
Formula:
C₈H₇N₃O₂
MDL Number:
MFCD00006890
:
M-Clarity™ quality level = MQ200
:
177.16 g/mol
EINECS:
208-309-4