Order Entry
Export
ContactUsLinkComponent
Luminol (3-Aminophthalhydrazide)
Luminol (3-Aminophthalhydrazide)
Catalog # A2185.0005
Supplier:  PanReac AppliChem
CAS Number:  
Luminol (3-Aminophthalhydrazide)
Catalog # A2185.0005
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :177.16 g/mol
EINECS:208-309-4
CAS Number:521-31-3
Formula:C₈H₇N₃O₂
MDL Number:MFCD00006890