Order Entry
Export
Orders LinkContactUsLinkComponent
Milk, powder

Milk, powder

Milk, powder
Supplier:  Roth Carl
Certificates

3 Options Available Below

Product Details & Documents
  • CAS Number:999999-99-4
  • EINECS:310-127-6

Documents

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 18,60 €
    18,60 €
    1 * 250 g
       18,60 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 28,60 €
    28,60 €
    1 * 500 g
       28,60 €

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 45,60 €
    45,60 €
    1 * 1 kg
       45,60 €