Order Entry
Export
Orders LinkContactUsLinkComponent
Milk, powder

Milk, powder

Glass bottle for solids, 20 g
Catalog # SIALBCR187-20G
Supplier:   Merck
Milk, powder
Catalog # SIALBCR187-20G
Supplier:  Merck
CAS Number:  999999-99-4
Milk, powder

Glass bottle for solids, 20 g

CAS Number:  999999-99-4
Price
324,00 €
Log in for availability
1 * 20 g
Product Details & Documents
  • CAS Number:999999-99-4
  • EINECS:310-127-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    20 g

Specifications