Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Pyridinedimethanol
2,6-Pyridinedimethanol, Sigma-Aldrich®
2,6-Pyridinedimethanol, Sigma-Aldrich®
2,6-Pyridinedimethanol, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
2,6-Pyridinedimethanol, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
1195-59-1
:
Formula:
C₇H₉NO₂
MDL Number:
MFCD00006351
:
139.15 g/mol
:
M-Clarity™ quality level = MQ100
EINECS:
214-803-0