Order Entry
Export
ContactUsLinkComponent
2,6-Pyridinedimethanol
2,6-Pyridinedimethanol
Catalog # MOLEM74966840
Supplier:  Molekula
CAS Number:  
2,6-Pyridinedimethanol
Catalog # MOLEM74966840
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :139.15 g/mol
EINECS:214-803-0
CAS Number:1195-59-1
Formula:C₇H₉NO₂
MDL Number:MFCD00006351