2,6-Pyridinedimethanol 98%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 2,6-Bis(hydroxymethyl)pyridine
- CAS Number:1195-59-1
- Formula:C₇H₉NO₂
- MDL Number:MFCD00006351
- Molecular Weight:139.15 g/mol
- Melting Pt:112…115 °C
- Storage Temperature:Ambient
- EINECS:214-803-0
3 Options Available Below
Product Details & Documents
Synonyms: 2,6-Bis(hydroxymethyl)pyridine
- CAS Number:1195-59-1
- Formula:C₇H₉NO₂
- MDL Number:MFCD00006351
- Molecular Weight:139.15 g/mol
- Melting Pt:112…115 °C
- Storage Temperature:Ambient
- EINECS:214-803-0
Documents
ACRO16755
SDS
Specifications for Product Family
Specification Test Results
- Appearance:White to yellow to beige Powder
- Infrared spectrum:Conforms
- Melting point:108 to 115 °C
- GC:≥97.5 %



