Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,4'-Dimethoxytrityl chloride
4,4'-Dimethoxytrityl chloride, Sigma-Aldrich®
4,4'-Dimethoxytrityl chloride, Sigma-Aldrich®
4,4'-Dimethoxytrityl chloride, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
4,4'-Dimethoxytrityl chloride, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
40615-36-9
:
Formula:
C₂₁H₁₉ClO₂
MDL Number:
MFCD00008409
:
338.8 g/mol
EINECS:
255-002-6