Order Entry
Export
ContactUsLinkComponent
4,4'-Dimethoxytrityl chloride
4,4'-Dimethoxytrityl chloride
Catalog # MOLE65276715-100G
Supplier:  Molekula
CAS Number:  
4,4'-Dimethoxytrityl chloride
Catalog # MOLE65276715-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :338.8 g/mol
EINECS:255-002-6
CAS Number:40615-36-9
Formula:C₂₁H₁₉ClO₂
MDL Number:MFCD00008409