Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Alcian blue
Alcian blue 8 GS (C.l. 74240)
Alcian blue 8 GS (C.l. 74240)
Irritant
Alcian blue 8 GS (C.l. 74240)
Supplier:
Roth Carl
Certificates
Expand image
Irritant
Alcian blue 8 GS (C.l. 74240)
Supplier:
Roth Carl
About this item
CAS Number:
33864-99-2
:
75881-23-1
Formula:
C₅₆H₆₈Cl₄CuN₁₆S₄
MDL Number:
MFCD00010720
:
1298.86 g/mol
EINECS:
251-705-7
:
GHS07
Storage Temperature:
Ambient