Ethyl-1,3-dithiane-2-carboxylate, Sigma-Aldrich®
Catalog # SIAL43749-5ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
:192.30 g/mol
EINECS:243-838-4
CAS Number:20462-00-4
Formula:C₇H₁₂O₂S₂
MDL Number:MFCD00006657