4-(Bromomethyl)tetrahydropyran
Catalog # MAYBCC29908DA
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:179.0549 g/mol
CAS Number:125552-89-8
Formula:C₆H₁₁BrO
MDL Number:MFCD03788561