
Imidazole-4,5-dicarboxylic acid ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:570-22-9
:
Formula:C₅H₄N₂O₄
MDL Number:MFCD00005200
Melting Pt:∼ 283 °C (dec.)
Density:1.749
:156.10 g/mol
EINECS:209-327-5
:GHS07
Storage Temperature:Ambient