Order Entry
Export
ContactUsLinkComponent
Imidazole-4,5-dicarboxylic acid 99%
 
Imidazole-4,5-dicarboxylic acid 99%
  ACRO412250250
 :  
undefined
 
Imidazole-4,5-dicarboxylic acid 99%
  ACRO412250250
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Light yellow to yellow-brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration excess NaOH:
    ≥98.5 %
  • UV:
    at 233 nm A: ≤1
  • Color scale:
    ≤5 APHA
  • Stabilizer:
    Approx. 0.4 % Methanol
  • Residue after evaporation:
    ≤0.0002 %
  • Acidity:
    ≤0.0001 meq/g
  • Filtered to 0.2 micrometer:
    Filtered
  • Free chlorine:
    ≤0.0005 %

 

 

 :
 :265 °C
 :156.10 g/mol
 :209-327-5
 :GHS07
 :Ambient
 :570-22-9
 :C₅H₄N₂O₄
 :MFCD00005200