Order Entry
Export
ContactUsLinkComponent
5-Fluoro-2'-deoxyuridine (floxuridine)
5-Fluoro-2'-deoxyuridine (floxuridine)
Catalog # MOLE27134875-1G
Supplier:  Molekula
CAS Number:  
5-Fluoro-2'-deoxyuridine (floxuridine)
Catalog # MOLE27134875-1G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :246.19 g/mol
EINECS:200-072-5
UN:2811
ADR:6.1,III
CAS Number:50-91-9
Formula:C₉H₁₁FN₂O₅
MDL Number:MFCD00006530