
2,2-Dimethylsuccinic anhydride 98%
:17347-61-4
:
:C₆H₈O₃
:MFCD00022608
:128.1259 g/mol
:219…220 °C
:26…30 °C
:108 °C
:Ambient
:241-371-0
:GHS07
- Appearance:White Low melting solid
- Infrared spectrum:Conforms
- Melting point:26 to 30 °C
- GC:≥97.5 %