Order Entry
Export
Orders LinkContactUsLinkComponent
(4-Methoxyphenoxy)acetonitrile

(4-Methoxyphenoxy)acetonitrile

Supplier:   Thermo Scientific
(4-Methoxyphenoxy)acetonitrile
Supplier:  Thermo Scientific
Certificates
(4-Methoxyphenoxy)acetonitrile

3 Options Available Below

Product Details & Documents
  • CAS Number:22446-12-4
  • Formula:C₉H₉NO₂
  • MDL Number:MFCD00017528
  • Molecular Weight:163.18 g/mol
  • UN:3276
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g