Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Difluoroacetone ≥98%

1,3-Difluoroacetone ≥98%

Toxic
Flammable
1,3-Difluoroacetone ≥98%
Certificates
1,3-Difluoroacetone ≥98%
1,3-Difluoroacetone ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 453-14-5 Molecular Formula: C3H4F2O Formula Weight: 94.06 Boiling Point: 101-102° UN No.: 2929 MDL No.: MFCD07368665

  • CAS Number:453-14-5
  • Formula:C₃H₄F₂O
  • MDL Number:MFCD07368665
  • Molecular Weight:94.06 g/mol
  • Boiling Pt:101…102 °C
  • Storage Temperature:Ambient
  • UN:2929
  • ADR:6.1,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 109,00 €
    109,00 €
    1 * 1 g
       109,00 €

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 404,00 €
    404,00 €
    1 * 5 g
       404,00 €