Order Entry
Export
ContactUsLinkComponent
Isopropyltriphenylphosphonium iodide, Sigma-Aldrich®
Isopropyltriphenylphosphonium iodide, Sigma-Aldrich®
Catalog # SIAL377481-25G
Supplier:  Merck
CAS Number:  
Isopropyltriphenylphosphonium iodide, Sigma-Aldrich®
Catalog # SIAL377481-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :432.29 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:246-281-5
CAS Number:24470-78-8
Formula:C₂₁H₂₂IP
MDL Number:MFCD00031595