

3,4-Dichlorobenzyl mercaptan ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:36480-40-7
:
Formula:C₇H₆Cl₂S
MDL Number:MFCD00039651
:193.10 g/mol
Boiling Pt:152…153 °C (12 mmHg)
Density:1.362
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:253-054-4
UN:2810
ADR:6.1,III
:GHS06
:GHS07